missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Certified AR for Analysis, Powder, Fisher Chemical™

Product Code. 10408210 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10408210 500 g 500g
10223180 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10408210 Supplier Fisher Chemical Supplier No. A/3680/53
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
MDL Number 10890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
IUPAC Name carbonic acid diamine
SMILES N.N.OC(O)=O

Specifications

Melting Point 58°C
Color White
pH 9.4
Physical Form Solid
Quantity 500 g
Formula Weight 96.09g/mol
Packaging Plastic powder jar
Vapor Pressure 69 hPa at 20°C
compliance-icons
Product Identifier
  • Ammonium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.