missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Extra Pure, Powder, SLR, Fisher Chemical™

Product Code. 10080030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
5kg
Product Code. Quantity Packaging unitSize
10080030 1 kg Plastic powder jar 1kg
10385260 5 kg Plastic keg 5kg
2 options
This item is not returnable. View return policy

Product Code. 10080030

Brand: Fisher Chemical A/3640/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
MDL Number 10890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
IUPAC Name carbonic acid diamine
SMILES N.N.OC(O)=O

Specifications

Melting Point 58°C
Color White
pH 9.4
Physical Form Solid
Quantity 1 kg
Formula Weight 96.09g/mol
Packaging Plastic powder jar
Vapor Pressure 69 hPa at 20°C
compliance-icons
Product Identifier
  • Ammonium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.