missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, NH3 ca 30%

Product Code. 11471698
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471698 1000 g 1000g
11481698 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471698 Supplier Thermo Scientific Alfa Aesar Supplier No. A13955.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol

Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Melting Point 60°C (sublimation)
Quantity 1000 g
Linear Formula (NH4)2CO3
Beilstein 3627235
Merck Index 14,508
Solubility Information Soluble in water.
Formula Weight 96.09
Odor Ammonia-like
Assay NH3 ≈30%
Chemical Name or Material Ammonium carbonate
compliance-icons
Product Identifier
  • Ammonium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.