missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, NH3 ca 30%

Product Code. 11481698
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
Product Code. Quantity unitSize
11481698 5000 g 5000g
11471698 1000 g 1000g
2 options
This item is not returnable. View return policy

Product Code. 11481698

Brand: Thermo Scientific Alfa Aesar A13955.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol

Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Melting Point 60°C (sublimation)
Quantity 5000 g
Linear Formula (NH4)2CO3
Beilstein 3627235
Merck Index 14,508
Solubility Information Soluble in water.
Formula Weight 96.09
Odor Ammonia-like
Assay NH3 ≈30%
Chemical Name or Material Ammonium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.