missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, puriss., Solstice
BIOPHARMA

Product Code. 15691020 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15691020 2.5 kg 2.5kg
15681020 1 kg 1kg
15601030 6 x 1 kg Pack of 6
15611030 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15691020 Supplier Honeywell-Fluka Supplier No. 1120425KG
 Restricted Item
Estimated Shipment: 16-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of NF, Ph. Franc., FCC

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Physical Form Crystalline
Quantity 2.5 kg
Linear Formula (NH4)2CO3
Beilstein 4253487
Formula Weight 96.09g/mol
Grade puriss.
Residue after Ignition ≤0.05% (as SO4)
Density Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium Carbonate, Meets analytical specification of NF, Ph.Franc.,FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.