missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, Puriss., meets analytical specification of NF, Ph. Franc., FCC, Solstice
BIOPHARMA

Product Code. 16665733
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16665733 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16665733 Supplier Fluka Supplier No. 1120425KG
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured

TRUSTED_SUSTAINABILITY

Specifications

CAS 1111-78-0, 1066-33-7
Quantity 25 kg
Molecular Formula CH8N2O3
InChI Key SFWLALIEAXEULE-UHFFFAOYSA-N
SMILES [NH4+].O.NC([O-])=O
IUPAC Name ammonium hydrate carbamate
Molecular Weight (g/mol) 96.09
Chemical Name or Material Ammonium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.