missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Cerium(Iv) Nitrate, ACS Reagent, ≥98.5%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
500g
50g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 214°C |
| Color | Orange |
| pH | Acidic |
| Quantity | 50 g |
| Linear Formula | Ce(NH4)2(NO3)6 |
| UN Number | UN3085 |
| Formula Weight | 548.22g/mol |
| Percent Purity | ≥98.5% |
| Grade | ACS Reagent |
| Packaging | Poly Bottle |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction