missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(IV) Nitrate, ge98% (titration), Honeywell™ Fluka™

Product Code. 15627750 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
Packaging:
Poly bottle
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 15627750

Brand: Honeywell-Fluka C3654250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥98% (titration)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 214°C
CAS 16774-21-3
Color Orange
pH Acidic
Physical Form Crystalline
Quantity 250 g
Linear Formula Ce(NH4)2(NO3)6
MDL Number MFCD00151121
UN Number UN3085
Solubility Information Soluble in water
Formula Weight 548.22g/mol
Percent Purity ≥98% (titration)
Odor Stinging
Packaging Poly bottle
Density 2.49g/cu.cm at 20°C
Chemical Name or Material Ammonium cerium(IV) nitrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.