missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(IV) Sulfate, Certified AR for analysis, low in rare earths, Fisher Chemical

Product Code. 10408400 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10408400 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10408400 Supplier Fisher Chemical Supplier No. A/3771/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 130°C
CAS 10378-47-9
Color Orange
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula CeH20N4O18S4
MDL Number 149417
InChI Key VCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILES N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
IUPAC Name λ4-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Molecular Weight (g/mol) 632.53
Formula Weight 632.54g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium cerium(IV) sulfate dihydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.