missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(Iv) Sulfate Dihydrate, Honeywell Fluka™

Product Code. 15653290 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
50 kg
Unit Size:
100g
50kg
Product Code. Quantity unitSize
15653290 50 kg 50kg
15643290 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 15653290

Brand: Honeywell-Fluka 3117350KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
MDL Number MFCD00149417

Specifications

Quantity 50 kg
Linear Formula Ce(NH4)4(SO4)4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 632.55g/mol
Percent Purity ≥99%
Chemical Name or Material Ammonium Cerium(Iv) Sulfate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.