missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cerium(IV) sulfate dihydrate, Honeywell Fluka™

Product Code. 15645300 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15645300 500 g 500g
15635300 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15645300 Supplier Honeywell-Fluka Supplier No. 383090500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥94%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
MDL Number MFCD00149417

Specifications

Quantity 500 g
Linear Formula Ce(NH4)4(SO4)4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 632.55g/mol
Percent Purity ≥94%
Chemical Name or Material Ammonium cerium(IV) sulfate dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.