missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, 99.5%, extra pure

Product Code. 10296920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
Plastic drum
Unit Size:
1kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10296920 1 kg Plastic bottle 1kg
10676052 500 g Plastic bottle 500g
10389133 5 kg Plastic drum 5kg
10135403 25 kg Fiber Drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10296920 Supplier Thermo Scientific Acros Supplier No. 199970010
  • ONLINE EXCLUSIVE PRICE
    73.65 €
    71.50 € / 1kg
    Save 2.15 €
    2% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12125-02-9 | ClH4N | 53.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206

Specifications

Melting Point 340.0°C
Color White
pH 4.5 to 5.5 (5% soln. at 20°C)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.5% min. (Argentometry)
Linear Formula NH3·HCl
Fieser 15,13
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate
SMILES N.Cl
IUPAC Name azanium;chloride
Formula Weight 53.49
Percent Purity 99.5%
Grade Extra Pure
Packaging Plastic bottle
Density 1.5200g/mL
Chemical Name or Material Ammonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.