missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10060030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
2kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10060030 500 g Plastic powder jar 500g
10213900 2 kg Plastic powder jar 2kg
10151750 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10060030 Supplier Fisher Chemical Supplier No. A/3920/53
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

H4ClN, CAS Number-12125-02-9, 5kg, White, 53.49g/mol, 340 deg.C, 11420, Plastic keg, Solid, 1.3mbar 160 | CAS: 12125-02-9 | ClH4N | 53.49 g/mol

H4ClN, CAS Number-12125-02-9, 5kg, White, 53.49g/mol, 340 deg.C, 11420, Plastic keg, Solid, 1.3mbar 160

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
IUPAC Name amine hydrochloride
SMILES N.Cl

Spécification

Melting Point 340°C
Color White
pH 5
Physical Form Solid
Quantity 500 g
Formula Weight 53.49g/mol
Packaging Plastic powder jar
Vapor Pressure 1.3mbar 160
compliance-icons
Identifiant produit
  • Ammonium chloride
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Mention de danger
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.