missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical

Product Code. 10070030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10070030 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10070030 Supplier Fisher Chemical Supplier No. A/3920/60
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 340°C
CAS 12125-02-9
Color White
pH 5
Physical Form Solid
Quantity 1 kg
Molecular Formula ClH4N
MDL Number MFCD00011420
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name azanium;chloride
Molecular Weight (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Formula Weight 53.49g/mol
Packaging Plastic powder jar
Vapor Pressure 1.3mbar 160
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.