Learn More
Ammonium chloride, Puratronic™, 99.999% (metals basis)
Description
Ammonium chloride finds its use as a flux in preparing metals for galvanizing, soldering or tin coating. While mainly referred as a nitrogen source fertilizer, it is also used in cleaning tip of soldering iron, hair shampoo as a thickening agent, some batteries as electrolyte, tanning, textile printing, dyeing, and cleaning products. In laboratories it is used to produce cooling baths and for producing buffers. Along with certain metals, it is used for effecting certain reductions in organic synthesis. It can be used as a precursor to ammonium cyanate, which is employed in the Wöhler synthesis of urea. It facilitates several organic transformations in a mild and efficient manner - for example in Ulmann coupling, cross-aldol condensation, Claisen rearrangement, reduction, thia-Michael addition and synthesis of several monocyclic, fused or spiro bicyclic nitrogen heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 340° (sublimation) |
| Color | White |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 100.00% |
| Solubility Information | Highly soluble in water. Soluble in liquid ammonia,acetone,hydrazine,alcohol. Insoluble in diethyl ether and ethyl acetate. |
| IUPAC Name | amine hydrochloride |
| Formula Weight | 53.49 |
| Odor | Odorless |
| Refractive Index | 1.642 |
| Show More |
Safety and Handling
- Ammonium chloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contain: Ammonium chloride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.