missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-100.5% (calc. to the dried substance), Solstice
BIOPHARMA

Product Code. 15621030
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
25kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15621030 1 kg 1kg
15631030 2.5 kg 2.5kg
15651030 6 x 1 kg Pack of 6
15641030 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15621030 Supplier Honeywell-Fluka Supplier No. 112091KG
  • ONLINE EXCLUSIVE PRICE
    84.00 €
    19.27 € / 1kg
    Save 64.73 €
    77% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name azanium;chloride

Specifications

Physical Form Solid
Quantity 1 kg
Linear Formula NH4Cl
Beilstein 4371014
SMILES N.Cl
Formula Weight 53.49g/mol
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Grade puriss.
Residue after Ignition ≤0.1% (as SO4)
Vapor Pressure 1mmHg (160.4°C)
Density Vapor Density: 1.9 (vs air)
Chemical Name or Material Ammonium Chloride, Meets analytical specification of Ph. Eur.BP, USP, FCC
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ammonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.