missing translation for 'onlineSavingsMsg'
Learn More

Ammonium citrate dibasic, Honeywell™

Product Code. 16615783 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Unit Size:
1kg
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16615783 1 kg 1kg
16625783 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16615783 Supplier Honeywell Research Chemicals Supplier No. 25102H1KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., ≥98%

  • Base grade products, suitability tested for specific life science applications such as molecular biology or cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3012-65-5
PubChem CID 13710713
ChEBI CHEBI:63076

Specifications

pH 4.5 - 5.5
Physical Form Solid
Quantity 1 kg
Percent Purity ≥98%
Grade Reagent
Packaging Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.