missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Ammonium Cuvette Tests

Product Code. 10000814
Click to view available options
Range:
2 to 47mg/L NH4-N
47 to 130mg/L NH4-N
Unit Size:
0.30kg
Each
This item is not returnable. View return policy

Product Code. 10000814

Brand: Hach Lange™ LCK303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7150-1, DIN 38406 E5-1
No. of Tests 25
Range 2 to 47mg/L NH4-N
Storage Requirements 2°C to 8°C
Test Method Indophenol Blue
Type Ammonium Cuvette Test
compliance-icons
Product Identifier
  • Ammonium, Sample cuvette
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium salicylate, Sodium hydroxide; caustic soda
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.