missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Ammonium Cuvette Tests

Product Code. 10566844
Change view
Click to view available options
Range:
2 to 47mg/L NH4-N
47 to 130mg/L NH4-N
Unit Size:
0.30kg
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10566844 47 to 130mg/L NH4-N Each
10000814 2 to 47mg/L NH4-N Each
10767572 2 to 47mg/L NH4-N 0.30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10566844 Supplier Hach Lange™ Supplier No. LCK302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7150-1, DIN 38406 E5-1
No. of Tests 25
Range 47 to 130mg/L NH4-N
Storage Requirements 2°C to 8°C
Test Method Indophenol Blue
Type Ammonium Cuvette Test
compliance-icons compliance-icons compliance-icons
Product Identifier
  • LatoN, Sample cuvette
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric acid, Phosphoric acid, Sodiumhydroxide, Potassiumnitrate, dipotassium peroxodisulphate,
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.