Learn More
Ammonium dichromate, ACS, 99.5% min, Thermo Scientific™
Catalysts, source of pure nitrogen, in lithography
90.30 € - 306.00 €
Chemical Identifiers
| CAS | 7789-09-5 |
|---|---|
| Molecular Formula | Cr2H8N2O7 |
| Molecular Weight (g/mol) | 252.06 |
| MDL Number | MFCD00010879 |
| InChI Key | JOSWYUNQBRPBDN-UHFFFAOYSA-P |
| Synonym | ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 |
| PubChem CID | 24600 |
| IUPAC Name | diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
| SMILES | [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11396788
|
Thermo Scientific Alfa Aesar
013444.36 |
500g |
90.30 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11386788
|
Thermo Scientific Alfa Aesar
013444.A3 |
2kg |
306.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| Cr2H8N2O7 | |
| MFCD00010879 | |
| ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
| diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Specifications
| 7789-09-5 | |
| Cr2H8N2O7 | |
| ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
| JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
| diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
| 24600 | |
| ACS Reagent |
| 99.5% | |
| MFCD00010879 | |
| Freely soluble in water.Soluble in water and alcohol. Insoluble in acetone. | |
| [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] | |
| 252.06 | |
| 252.1 | |
| Ammonium dichromate |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H272-H301-H312-H314-H317-H330-H334-H340-H350-H360FD-H372-H500
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 232-143-1
missing translation for 'rtecsNumber' : HX7650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.