missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Dichromate, TRC

Product Code. 30329412
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329412 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329412 Supplier Toronto Research Chemicals Supplier No. TRCA63398050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium dichromate
CAS 7789-09-5
Molecular Formula Cr2 O7 . 2 H4 N
Quantity 50 g
Synonym Chromic acid (H2Cr2O7), ammonium salt (1:2), Chromic acid (H2Cr2O7), diammonium salt (9CI), Dichromic acid (H2Cr2O7), diammonium salt (8CI), Ammonium bichromate, Ammonium chromate ((NH4)2Cr2O7), Ammonium dichromate, Ammonium dichromate(VI), Diammonium dichromate
InChI Formula InChI=1S/2Cr.2H3N.7O/h;;2*1H3;;;;;;;/q;;;;;;;;;2*-1/p+2
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 252.06
Formula Weight 251.9142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.