missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 99%, for HPLC
GREENER_CHOICE

Product Code. 12635087 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12635087 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12635087 Supplier Thermo Scientific Supplier No. 447382500
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 190.0°C
CAS 7722-76-1
Quantity 250 g
Packaging Plastic bottle
Assay Percent Range 99%
Molecular Formula H6NO4P
Linear Formula NH4H2PO4
MDL Number MFCD00003396
Merck Index 15,541
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
Solubility Information (10% in water) Clear colorless
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES N.OP(O)(O)=O
IUPAC Name phosphoric acid amine
Molecular Weight (g/mol) 115.03
PubChem CID 24402
ChEBI CHEBI:62982
Formula Weight 115.03
Percent Purity 99%
Grade HPLC
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.