missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, ACS, 98.0% min

Product Code. 11307347
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307347 500 g 500g
11317347 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307347 Supplier Thermo Scientific Alfa Aesar Supplier No. 011598.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol

Preparation of baking powder with sodium bicarbonate, in fermentation.Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98% min.
Solubility Information Freely soluble in water. Slightly soluble in alcohol. Insoluble in acetone
Formula Weight 115.03
Grade ACS
Odor Odorless
Density 1.803 g/cm3
Chemical Name or Material Ammonium dihydrogen phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.