missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 0.5M solution in methanol, Thermo Scientific™

Product Code. 10793614 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
100mL; Plastic bottle
This item is not returnable. View return policy

Product Code. 10793614

Brand: Thermo Scientific Acros 393011000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 12125-01-8, 67-56-1
Quantity 100mL
Molecular Formula H4FN
Linear Formula NH4F
MDL Number MFCD00011423
Flash Point 11°C
Merck Index 15, 519
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 37.04
Formula Weight 37.04
Packaging Plastic bottle
Specific Gravity 0.82
Density 0.8200g/mL
Chemical Name or Material Ammonium fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium fluoride, 0.5M solution in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.