missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 96%

Product Code. 11467737
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11467737

Brand: Thermo Scientific Alfa Aesar A16279.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12125-01-8 | FH4N | 37.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.

Solubility
Freely soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic and non-cumbustible. On heating, it sublimes and decomposes to hydrogen fluoride and ammonia gas, which can recombine to form ammonium fluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Quantity 100 g
Linear Formula NH4F
UN Number UN2505
Merck Index 14,522
Solubility Information Freely soluble in water. Slightly soluble in alcohol.
Formula Weight 37.04
Percent Purity 96%
Odor Odorless
Sensitivity Hygroscopic
Density 1.01
Chemical Name or Material Ammonium fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.