missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, Puriss. p.a., ACS Reagent, ≥98%, Honeywell Fluka™

Product Code. 15672970 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
1 kg
50 kg
6 x 1 kg
Unit Size:
100g
1kg
50kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15672970

Brand: Honeywell-Fluka 301011KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Quantity 1 kg
Linear Formula NH4F
UN Number 2505
Formula Weight 37.04g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Packaging Plastic Bottle
Residue after Ignition ≤0.005% (as SO4)
Chemical Name or Material Ammonium Fluoride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.