missing translation for 'onlineSavingsMsg'
Learn More

Ammonium formate, 97%

Product Code. 10164180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10164180 500 g Glass bottle 500g
10205650 1 kg Plastic bottle 1kg
11914891 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164180 Supplier Thermo Scientific Acros Supplier No. 168615000
  • ONLINE EXCLUSIVE PRICE
    44.40 €
    41.60 € / 500g
    Save 2.80 €
    6% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 540-69-2 | CH5NO2 | 63.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.06
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
PubChem CID 2723923
ChEBI CHEBI:63050
IUPAC Name azanium;formate
SMILES C(=O)[O-].[NH4+]

Specifikationer

Melting Point 115.0°C to 120.0°C
Color White
pH 6 to 7 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula HCO2NH4
Beilstein 02,8
Fieser 01,38; 13,230; 15,13
Merck Index 15,52
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 63.06
Percent Purity 97%
Chemical Name or Material Ammonium formate
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Ammonium formate
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.