missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium formate, 99%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 115.0°C to 120.0°C |
| Color | White |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 5 kg |
| Assay Percent Range | 99% |
| Linear Formula | HCO2NH4 |
| Beilstein | 02,8 |
| Fieser | 01,38; 13,230; 15,13 |
| Merck Index | 15,52 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol |
| Show More |
Safety and Handling
Product Identifier
- Ammonium formate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction