missing translation for 'onlineSavingsMsg'
Learn More

Ammonium formate, for LC-MS, Solstice

Product Code. 15635830
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15635830 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15635830 Supplier Honeywell-Fluka Supplier No. 5567450G
  • ONLINE EXCLUSIVE PRICE
    231.25 €
    219.14 € / 50g
    Save 12.11 €
    5% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 540-69-2
Physical Form Crystalline
Quantity 50 g
Molecular Formula CH5NO2
Linear Formula HCO2NH4
MDL Number MFCD00013103
UN Number NONH for all modes of transport
Beilstein 3625095
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
SMILES C(=O)[O-].[NH4+]
IUPAC Name azanium;formate
Molecular Weight (g/mol) 63.056
PubChem CID 2723923
ChEBI CHEBI:63050
Formula Weight 63.06g/mol
Percent Purity ≥99% (calculated on dry substance, NT)
Density 1.26g/mL at 25°C
Chemical Name or Material Ammonium formate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium formate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.