missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, LC-MS Ultra, Solstice

Product Code. 15681400
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15681400 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15681400 Supplier Honeywell-Fluka Supplier No. 1426625G
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™ LC-MS Ultra; eluent additive for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles
TRUSTED_SUSTAINABILITY

Specifications

CAS 540-69-2
Physical Form Crystalline Powder
Quantity 25 g
Molecular Formula CH5NO2
Linear Formula HCO2NH4
MDL Number MFCD00013103
UN Number NONH for all modes of transport
Beilstein 3625095
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
SMILES C(=O)[O-].[NH4+]
IUPAC Name azanium;formate
Molecular Weight (g/mol) 63.056
PubChem CID 2723923
ChEBI CHEBI:63050
Formula Weight 63.06g/mol
Grade LC-MS Ultra
Density 1.26g/mL at 25°C
Chemical Name or Material Ammonium Formate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium formate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.