missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, Reagent grade, 97%, Honeywell Burdick & Jackson™

Product Code. 15671500 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15671500 500 g Poly Bottle 500g
15641500 100 g Poly Bottle 100g
15651500 1 kg Poly Bottle 1kg
15661500 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15671500 Supplier Honeywell Research Chemicals Supplier No. 156264500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.056
MDL Number MFCD00013103
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
PubChem CID 2723923
ChEBI CHEBI:63050
IUPAC Name azanium;formate
SMILES C(=O)[O-].[NH4+]

Specifications

Melting Point 116°C
Color White
pH 6.0 to 7.0 (at 20°C)
Quantity 500 g
Linear Formula HCO2NH4
UN Number NONH for all modes of transport
Beilstein 3625095
Formula Weight 63.06g/mol
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Appearance Crystals/Granules
Density 1.26g/mL at 25°C
Chemical Name or Material Ammonium Formate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.