missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Iridium complexes are generated from ammonium hexachloroiridate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | Black-Red |
| Physical Form | Crystalline Powder |
| Quantity | 5 g |
| Solubility Information | Soluble in hydrochloric acid,lightly soluble in water,insoluble in alcohol |
| IUPAC Name | ammonium hexachloroiridate |
| Formula Weight | 441.02 |
| Percent Purity | 99% |
| Concentration or Composition (by Analyte or Components) | Ir 43% min. |
| Sensitivity | Hygroscopic |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction