missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroosmate(IV), 99.9% (metals basis), Os 42.5% min

Product Code. 11320118
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11320118

missing translation for 'mfr': Thermo Scientific Alfa Aesar 042874.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 12125-08-5 | Cl6H8N2Os | 439.00 g/mol

Ammonium hexachloroosmate(IV) acts as a pharmaceutical, organic and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-08-5
Molecular Formula Cl6H8N2Os
Molecular Weight (g/mol) 439.00
MDL Number MFCD00010883
InChI Key JRHMYOOMZKAPKT-UHFFFAOYSA-J
Synonym ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
PubChem CID 11729867
IUPAC Name diazanium;hexachloroosmium(2-)
SMILES [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 170° (sublimation)
Color Brown to Black
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.9% (Os 42.5% min.) (Metals basis)
Solubility Information Soluble in water.
Formula Weight 438.99
Sensitivity Hygroscopic
Density 2.9
Chemical Name or Material Ammonium hexachloroosmate(IV)
compliance-icons
Product Identifier
  • Ammonium hexachloroosmiate(IV)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium hexachloroosmiate(IV)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.