missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloropalladate(IV), 99.9% (metals basis), Pd 29%

Product Code. 11361678
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361678 10 g 10g
11351678 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361678 Supplier Thermo Scientific Alfa Aesar Supplier No. 011042.09
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 19168-23-1 | Cl6H8N2Pd | 355.20 g/mol

Ammonium hexachloropalladate(IV) is used as analytical reagent and as a catalyst in the chemical synthesis. It is also used as raw material and intermediates in organic synthesis and pharmaceutical field. Further, it is used in emission spectrographic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19168-23-1
Molecular Formula Cl6H8N2Pd
Molecular Weight (g/mol) 355.20
MDL Number MFCD00015959
InChI Key AZUQNARGWZAFRM-UHFFFAOYSA-J
Synonym ammonium hexachloropalladate iv, ammonium hexachloropalladate, diammonium hexachloropalladate, diammonium hexachloropalladate 2-, ccris 7829, hexachloropalladate 2-diammonium, ammonium chloropalladate nh4 2pdcl6, palladate 2-, hexachloro-, diammonium, oc-6-11, palladate 2-, hexachloro-, diammonium, epitope id:151517
PubChem CID 62732
ChEBI CHEBI:60124
SMILES [NH4+].[NH4+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Red to Orange
Physical Form Crystalline
Quantity 10 g
UN Number UN3077
Solubility Information Slightly soluble in water.
IUPAC Name diazanium;hexachloropalladium(2-)
Formula Weight 355.2
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 29%
Odor Odorless
Sensitivity Hygroscopic
Density 2.42 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachloropalladate(IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.