missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorophosphate, 99%, pure

Product Code. 10616062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616062 100 g Plastic bottle 100g
10312523 5 g Plastic bottle 5g
10032730 25 g Plastic bottle 25g
10287403 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10616062 Supplier Thermo Scientific Acros Supplier No. 202321000
  • ONLINE EXCLUSIVE PRICE
    318.00 €
    298.00 € / 100g
    Save 20.00 €
    6% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16941-11-0 | H4F6NP | 163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16941-11-0
Molecular Formula H4F6NP
Molecular Weight (g/mol) 163
MDL Number MFCD00064642
InChI Key NIZXKAYXSNUDOU-UHFFFAOYSA-O
Synonym ammonium hexafluorophosphate, ammonium hexafluorophosphate v, ammonium hexafluophosphate, ammoniumhexafluorophosphate 6ci,7ci, unii-ns308031pj, azanium hexafluorophosphate, nh4pf6, ksc492g2j, ammonium hexafluoro-, e5-phosphanuide, ammonium phosphorus hexafluoride
PubChem CID 9793912
IUPAC Name azanium;hexafluorophosphate
SMILES [NH4+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point 198°C
Color White to Beige
Physical Form Crystalline Powder or Needles
Quantity 100 g
Packaging Plastic bottle
Specific Gravity 2.18
Assay Percent Range 99%
Density 2.1800g/mL
Linear Formula NH4PF6
Merck Index 15, 523
Solubility Information Solubility in water: 74.8g/100mL (20 c). Other solubilities: soluble in acetone,methanol,ethanol and methyl,acetate
Formula Weight 163
Percent Purity 99%
Grade Pure
Chemical Name or Material Ammonium hexafluorophosphate, 99.5%
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.