missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydrogen Difluoride, 95%, Reagent grade, Solstice

Product Code. 15662240 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Poly Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15662240 25 g Poly Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15662240 Supplier Honeywell Research Chemicals Supplier No. 22482025G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Reagent Grade, 95%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Melting Point 125°C
Color White
Quantity 25 g
Packaging Poly Bottle
Appearance Crystals or flakes
Density 1.500g/cm3 at 20°C
Linear Formula NH4HF2
UN Number UN1727
Formula Weight 57.04g/mol
Percent Purity 95%
Grade Reagent
Chemical Name or Material Ammonium Hydrogen Difluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.