missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydrogen Difluoride, Extra Pure, SLR, Fisher Chemical™

Product Code. 11433513 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433513 2.5 kg 2.5kg
12327133 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11433513 Supplier Fisher Chemical Supplier No. A/5280/62
  • ONLINE EXCLUSIVE PRICE
    165.00 €
    163.00 € / 2.5kg
    Save 2.00 €
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
IUPAC Name ammonium hydrogen difluoride
SMILES N.F.F

Specifications

Boiling Point 230°C
Melting Point 125°C
Color White
pH 3.5
Physical Form Solid
Quantity 2.5 kg
Formula Weight 57.04g/mol
Packaging Plastic powder jar
Vapor Pressure 1 hPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • Ammonium hydrogen difluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.