missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydrogen Difluoride, Honeywell Fluka™

Product Code. 15690910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15690910 50 g 50g
15680910 250 g 250g
15670910 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15690910 Supplier Honeywell-Fluka Supplier No. 0983550G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Puriss. p.a.,≥98.5% (F)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Quantity 50 g
Linear Formula NH4HF2
UN Number 1727
Formula Weight 57.04g/mol
Percent Purity ≥98.5% (F)
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Ammonium Hydrogen Difluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.