missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, 30%, J.T. Baker™

Product Code. 15538654 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15538654 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15538654 Supplier Avantor J.T.Baker Supplier No. 973303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
Boiling Point 37.7°C
CAS 1336-21-6
Melting Point -57.5°C
Molecular Formula H5NO
MDL Number MFCD00066650
Density 0.91g/cm3
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 4 L
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.04
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.