missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, for analysis, 20% solution in water

Product Code. 11300846 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11300846 2.5 L Glass Bottle 2.5 L
10300864 1 L Glass Bottle 1 L
10278993 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11300846 Supplier Thermo Scientific Acros Supplier No. 390070025
  • ONLINE EXCLUSIVE PRICE
    57.10 €
    55.40 € / 2.5 L
    Save 1.70 €
    2% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1336-21-6 | H5NO | 35.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Color Brown-Green to Brown-Purple
Linear Formula NH4OH
Density 0.9200g/mL
Merck Index 15, 491
Solubility Information Solubility in water: soluble.
Quantity 2.5 L
IUPAC Name azanium;hydroxide
Formula Weight 35.05
Percent Purity ≥20% (Acidimetry)
Grade Analytical
Packaging Glass Bottle
Specific Gravity 0.92
Physical Form Solution
Chemical Name or Material Ammonium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.