missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide solution, Puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis, Honeywell Fluka™
BIOPHARMA

Product Code. 15660780 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 kg
4 x 5 L
50 kg
6 x 1 L
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Plastic Pail
Unit Size:
1 L
2.5 L
25kg
5 L
50kg
Pack of 4
Pack of 6
Product Code. Quantity Packaging unitSize
15660780 25 kg Plastic Pail 25kg
15630780 1 L Plastic Bottle 1 L
15650780 2.5 L Glass Bottle 2.5 L
15640780 2.5 L Plastic Bottle 2.5 L
15600790 5 L Plastic Bottle 5 L
15610790 6 x 1 L Glass Bottle Pack of 6
15680780 4 x 5 L Plastic Bottle Pack of 4
15690780 50 kg Metal Drum 50kg
8 options
This item is not returnable. View return policy

Product Code. 15660780

Brand: Honeywell-Fluka 0500325KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Linear Formula NH4OH
Concentration 25 to 30% NH3 basis
UN Number 2672
Beilstein 3587154
Density 0.892 to 0.910g/mL at 20°C, 0.9g/mL at 25°C
Quantity 25 kg
IUPAC Name azanium;hydroxide
Explosion Limit 0.27
Formula Weight 35.05g/mol
Grade puriss.
Packaging Plastic Pail
Chemical Name or Material Ammonium Hydroxide Solution, Meets analytical specification of Ph.Eur.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide solutio
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonia, aqueous solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.