missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide Solution, puriss. p.a., 0%, Solstice

Product Code. 15670920
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15670920 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15670920 Supplier Honeywell-Fluka Supplier No. 098611L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Puriss. p.a., packed in coated, shock- and leak-protected glass bottle, 25% NH3 in H2O (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

pH Alkaline
Color Colorless
Boiling Point 24°C (1013hPa)
CAS 1336-21-6
Melting Point -92°C
Molecular Formula H5NO
Linear Formula NH4OH
MDL Number MFCD00066650
UN Number UN2672
Beilstein 3587154
Density 0.91g/mL at 20°C, 0.9g/mL at 25°C
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Solubility Information Completely miscible
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 1 L
SMILES N.O
IUPAC Name azanium;hydroxide
Explosion Limit 0.27
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.05g/mol
Percent Purity 0%
Grade puriss. p.a.
Odor Stinging ammoniacal
Packaging Glass bottle
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.