missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iodide, puriss., Solstice

Product Code. 15660660 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
50 kg
6 x 1 kg
Unit Size:
100g
1kg
50kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15660660 100 g 100g
15670660 1 kg 1kg
15690660 6 x 1 kg Pack of 6
15680660 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15660660 Supplier Honeywell-Fluka Supplier No. 03101100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
SMILES [NH4+].[I-]

Specifications

Quantity 100 g
Linear Formula NH4I
UN Number NONH for all modes of transport
IUPAC Name ammonium iodide
Formula Weight 144.94g/mol
Percent Purity ≥99%
Grade puriss.
Residue after Ignition ≤0.1% (as SO4)
Vapor Pressure 1mmHg (210.9°C)
Chemical Name or Material Ammonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.