Learn More
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Melting Point | 100.0°C |
| Color | Blue-Green |
| Physical Form | Crystals |
| Quantity | 100 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 99% |
| Linear Formula | (NH4)2Fe(SO4)2·6H2O |
| Merck Index | 15, 518 |
| Solubility Information | Solubility in water: 269g/L (20°C). Other solubilities: practically insoluble in alcohol |
| Formula Weight | 392.14 |
| Afficher plus |
Sécurité et traitement
- Ammonium iron (II) sulfate hexahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.