missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis

Product Code. 10532991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
25g
25kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10532991 500 g Plastic bottle 500g
10762241 25 g Plastic bottle 25g
10052750 1 kg Plastic bottle 1kg
10553124 5 kg Plastic bottle 5kg
10444985 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10532991 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 201375000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ferrous ammonium sulfate, Mohr's salt
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 100.0°C
Color Green
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula (NH4)2Fe(SO4)2·6H2O
Merck Index 15, 518
Solubility Information Solubility in water: 269g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 392.14
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium iron (II) sulfate hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.