missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(Ii) Sulfate Hexahydrate, puriss. p.a., Solstice

Product Code. 15670890
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15670890 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15670890 Supplier Honeywell-Fluka Supplier No. 097201KG
  • ONLINE EXCLUSIVE PRICE
    86.00 €
    75.67 € / 1kg
    Save 10.33 €
    12% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., Reagent ISO, Reagent European Pharmacopoeia,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-85-9
Quantity 1 kg
Molecular Formula FeH20N2O14S2
Linear Formula (NH4)2Fe(SO4)2 · 6H2O
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
Molecular Weight (g/mol) 392.13
Formula Weight 392.14g/mol
Percent Purity ≥99% (RT)
Grade puriss. p.a.
Chemical Name or Material Ammonium Iron(Ii) Sulfate Hexahydrate, Reag. Ph. Eur. reag. ISO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.