missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) sulfate hexahydrate, Puriss. p.a.,Reag ISO, Reag Eur Pharmacopoeia, ≥99.0% (RT), Solstice

Product Code. 16673944
Change view
Click to view available options
50 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16673944 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16673944 Supplier Fluka Supplier No. 0972050KG
  • ONLINE EXCLUSIVE PRICE
    3430.00 €
    3097.50 € / 50kg
    Save 332.50 €
    9% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 100°C (Decomposes before melting)
CAS 7783-85-9
Color Blue Green
CAS Max % 1
pH 3 to 5
Quantity 50 kg
Molecular Formula FeH20N2O14S2
Linear Formula (NH4)2Fe(SO4)2 · 6H2O
MDL Number MFCD00150530
Synonym Ammonium ferrous sulfate hexahydrate; Ammonium iron(II) sulfate; MohrAmmonium ferrous sulfate hexahydrate; Ammonium iron(II) sulfate; Mohr's salts salt
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name iron(3+) diammonium hexahydrate disulfate
Molecular Weight (g/mol) 392.13
Grade ACS
Density 1.86 g/cm3 (20°C)
Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.