missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11398668
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398668 1 g 1g
11308678 5 g 5g
11318678 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398668 Supplier Thermo Scientific Alfa Aesar Supplier No. 040751.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol

Ammonium iron(II) sulfate hydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Iron(II) ammonium sulfate; Ferrous ammonium sulfate
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point ∼100°C (decomposition)
Color Blue-Green
Physical Form Coarse Powder
Quantity 1 g
Odor Odorless
Sensitivity Air and light sensitive
Density 1.864 g/mL
Linear Formula (NH4)2Fe(SO4)2·xH2O (x≈6)
Merck Index 14,521
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 284.04 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Ammonium iron(II) sulfate hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium iron(II) sulfate hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.