missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron (II) Sulfate Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, Fisher Chemical™

Product Code. 10375732 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10375732 2.5 L HDPE plastic bottle 2.5 L
10060180 1 L Amber glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10375732 Supplier Fisher Chemical Supplier No. J/1101/PB17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Physical Form Liquid
Quantity 2.5 L
Packaging HDPE plastic bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.