missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(III) Citrate, TRC

Product Code. 30134298
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134298 500 g 500g
17979104 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134298 Supplier Toronto Research Chemicals Supplier No. TRCA634913500G
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H8FeNO7
Molecular Weight (g/mol) 261.98
Synonym Ferric Ammonium Citrate
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron
SMILES N.[Fe+3].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-]

Specifications

Quantity 500 g
InChI Formula InChI=1S/C6H8O7.Fe.H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;1H3
Formula Weight 261.965
Chemical Name or Material Ammonium Iron(III) Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.